Assay Details
| Gene Symbol: mnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003597.2 | NP_001003597.1 | 7 - 8 | 561 | 98 | |
| GenBank mRNA | BC078223.1 | - | 7 - 8 | 565 | 98 | |
More Information
Set Membership: |
|

mnd1
Genomic Map