Assay Details
| Gene Symbol: lypd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004670.1 | NP_001004670.1 | 4 - 5 | 540 | 73 | |
| XM_021478643.1 | XP_021334318.1 | 4 - 5 | 406 | 73 | ||
| GenBank mRNA | BC081426.1 | - | 4 - 5 | 540 | 73 | |
More Information
Set Membership: |
|

lypd6
Genomic Map