Assay Details
| Gene Symbol: fabp4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004682.1 | NP_001004682.1 | 2 - 3 | 311 | 93 | |
| GenBank mRNA | AY628221.2 | - | 2 - 3 | 314 | 93 | |
| BC081489.1 | - | 2 - 3 | 311 | 93 | ||
More Information
Set Membership: |
|

fabp4a
Genomic Map