Assay Details
| Gene Symbol: ank1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005969.2 | NP_001005969.2 | 1 - 2 | 408 | 58 | |
| XM_005165647.4 | XP_005165704.1 | 1 - 2 | 448 | 58 | ||
| XM_005165648.4 | XP_005165705.1 | 1 - 2 | 448 | 58 | ||
| XM_005165649.4 | XP_005165706.1 | 1 - 2 | 446 | 58 | ||
| XM_021476120.1 | XP_021331795.1 | 1 - 2 | 452 | 58 | ||
| GenBank mRNA | BC083296.1 | - | 1 - 2 | 402 | 58 | |
More Information
Set Membership: |
|

ank1a
Genomic Map