Assay Details
| Gene Symbol: tmem144b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005983.1 | NP_001005983.1 | 1 - 2 | 129 | 62 | |
| GenBank mRNA | BC083426.1 | - | 1 - 2 | 129 | 62 | |
More Information
Set Membership: |
|

tmem144b
Genomic Map