Assay Details
| Gene Symbol: elovl1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005989.3 | NP_001005989.1 | 3 - 4 | 561 | 71 | |
| XM_005171196.4 | XP_005171253.1 | 3 - 4 | 605 | 71 | ||
| GenBank mRNA | BC083417.1 | - | 3 - 4 | 317 | 71 | |
| EH495486.1 | - | 3 - 4 | 503 | 71 | ||
| EH549422.1 | - | 3 - 4 | 602 | 71 | ||
More Information
Set Membership: |
|

elovl1a
Genomic Map