Assay Details
| Gene Symbol: elovl1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005989.3 | NP_001005989.1 | 6 - 7 | 805 | 93 | |
| XM_005171196.4 | XP_005171253.1 | 6 - 7 | 849 | 93 | ||
| GenBank mRNA | BC083417.1 | - | 6 - 7 | 561 | 93 | |
| EH495486.1 | - | 6 - 7 | 748 | 94 | ||
| EH549422.1 | - | 6 - 7 | 846 | 93 | ||
More Information
Set Membership: |
|

elovl1a
Genomic Map