Assay Details
| Gene Symbol: elovl1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005989.3 | NP_001005989.1 | 7 - 8 | 936 | 80 | |
| XM_005171196.4 | XP_005171253.1 | 7 - 8 | 980 | 80 | ||
| GenBank mRNA | BC083417.1 | - | 7 - 8 | 692 | 80 | |
| EH570542.1 | - | 1 - 2 | 688 | 80 | ||
More Information
Set Membership: |
|

elovl1a
Genomic Map