Assay Details
| Gene Symbol: tprkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007373.1 | NP_001007374.1 | 2 - 3 | 347 | 140 | |
| XM_021475009.1 | XP_021330684.1 | 2 - 3 | 348 | 140 | ||
| GenBank mRNA | BC085466.1 | - | 2 - 3 | 347 | 140 | |
More Information
Set Membership: |
|

tprkb
Genomic Map