Assay Details
| Gene Symbol: bin2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007421.1 | NP_001007422.1 | 5 - 6 | 471 | 84 | |
| XM_009296976.3 | XP_009295251.1 | 5 - 6 | 565 | 84 | ||
| XM_009296977.3 | XP_009295252.1 | 5 - 6 | 618 | 84 | ||
| GenBank mRNA | BC085363.1 | - | 5 - 6 | 471 | 84 | |
| BC165739.1 | - | 4 - 5 | 358 | 84 | ||
More Information
Set Membership: |
|

bin2a
Genomic Map