Assay Details
| Gene Symbol: nap1l4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007453.1 | NP_001007454.1 | 9 - 10 | 909 | 86 | |
| XM_009303210.3 | XP_009301485.1 | 9 - 10 | 986 | 86 | ||
| XM_021477517.1 | XP_021333192.1 | 8 - 9 | 923 | 86 | ||
| GenBank mRNA | BC085386.1 | - | 9 - 10 | 909 | 86 | |
| BC164768.1 | - | 8 - 9 | 874 | 86 | ||
More Information
Set Membership: |
|

nap1l4b
Genomic Map