Assay Details
| Gene Symbol: rasd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007781.2 | NP_001007782.1 | 1 | 555 | 65 | |
| GenBank mRNA | AY729884.2 | - | 1 | 837 | 65 | |
| BC124474.1 | - | 1 | 555 | 65 | ||
More Information
Set Membership: |
|

rasd4
Genomic Map