Assay Details
| Gene Symbol: gna13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013263.2 | NP_001013281.2 | 1 - 2 | 306 | 85 | |
| GenBank mRNA | AY386361.1 | - | 1 - 2 | 284 | 85 | |
| BC134004.1 | - | 1 - 2 | 306 | 85 | ||
More Information
Set Membership: |
|

gna13b
Genomic Map