Assay Details
| Gene Symbol: gna13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013263.2 | NP_001013281.2 | 3 - 4 | 586 | 95 | |
| GenBank mRNA | AY386361.1 | - | 3 - 4 | 564 | 95 | |
| BC134004.1 | - | 3 - 4 | 586 | 95 | ||
More Information
Set Membership: |
|

gna13b
Genomic Map