Assay Details
| Gene Symbol: arpin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017780.1 | NP_001017780.1 | 1 - 2 | 129 | 80 | |
| GenBank mRNA | BC093306.1 | - | 1 - 2 | 129 | 80 | |
More Information
Set Membership: |
|

arpin
Genomic Map