Assay Details
| Gene Symbol: bdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017809.1 | NP_001017809.1 | 8 - 9 | 637 | 75 | |
| XM_005160067.4 | XP_005160124.1 | 8 - 9 | 665 | 75 | ||
| XM_005160068.4 | XP_005160125.1 | 8 - 9 | 906 | 75 | ||
| GenBank mRNA | BC092874.1 | - | 8 - 9 | 637 | 75 | |
| BC165437.1 | - | 7 - 8 | 623 | 75 | ||
More Information
Set Membership: |
|

bdh2
Genomic Map