Assay Details
| Gene Symbol: slc18b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017841.1 | NP_001017841.1 | 8 - 9 | 988 | 74 | |
| XM_005158600.4 | XP_005158657.1 | 9 - 10 | 1042 | 74 | ||
| GenBank mRNA | BC092773.1 | - | 8 - 9 | 988 | 74 | |
More Information
Set Membership: |
|

slc18b1
Genomic Map