Assay Details
| Gene Symbol: endou2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020562.1 | NP_001018398.1 | 2 - 3 | 202 | 67 | |
| XM_009295679.3 | XP_009293954.1 | 2 - 3 | 274 | 67 | ||
| GenBank mRNA | BC095160.1 | - | 2 - 3 | 202 | 67 | |
| BC164551.1 | - | 1 - 2 | 132 | 67 | ||
More Information
Set Membership: |
|

endou2
Genomic Map