Assay Details
| Gene Symbol: gba3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020693.1 | NP_001018529.1 | 3 - 4 | 1171 | 100 | |
| GenBank mRNA | BC095794.1 | - | 3 - 4 | 1171 | 100 | |
| BC165748.1 | - | 3 - 4 | 1132 | 100 | ||
More Information
Set Membership: |
|

gba3
Genomic Map