Assay Details
| Gene Symbol: gfi1aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020776.1 | NP_001018612.1 | 3 - 4 | 426 | 62 | |
| GenBank mRNA | BC095882.1 | - | 3 - 4 | 426 | 62 | |
| BC165554.1 | - | 2 - 3 | 328 | 62 | ||
More Information
Set Membership: |
|

gfi1aa
Genomic Map