Assay Details
| Gene Symbol: gfi1aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020776.1 | NP_001018612.1 | 5 - 6 | 939 | 129 | |
| GenBank mRNA | BC095882.1 | - | 5 - 6 | 939 | 129 | |
| BC165554.1 | - | 4 - 5 | 841 | 129 | ||
More Information
Set Membership: |
|

gfi1aa
Genomic Map