Assay Details
| Gene Symbol: metap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025165.2 | NP_001020336.2 | 1 - 2 | 179 | 69 | |
| XM_017358739.2 | XP_017214228.2 | 1 - 2 | 157 | 69 | ||
| GenBank mRNA | BC076042.1 | - | 1 - 2 | 94 | 69 | |
| BC096796.1 | - | 135 | 69 | |||
| EH501151.1 | - | 1 - 2 | 169 | 69 | ||
More Information
Set Membership: |
|

metap1
Genomic Map