Assay Details
| Gene Symbol: herpud1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025537.1 | NP_001020708.1 | 6 - 7 | 1055 | 66 | |
| XM_005159120.4 | XP_005159177.1 | 6 - 7 | 1158 | 66 | ||
| XM_005159121.4 | XP_005159178.1 | 6 - 7 | 1111 | 66 | ||
| GenBank mRNA | BC097190.1 | - | 6 - 7 | 1055 | 66 | |
More Information
Set Membership: |
|

herpud1
Genomic Map