Assay Details
| Gene Symbol: zgc:101731 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025558.2 | NP_001020729.1 | 6 - 7 | 563 | 80 | |
| XM_005157730.4 | XP_005157787.1 | 6 - 7 | 566 | 80 | ||
| XM_005157731.4 | XP_005157788.1 | 6 - 7 | 561 | 80 | ||
| XM_005157732.4 | XP_005157789.1 | 7 - 8 | 674 | 80 | ||
| GenBank mRNA | BC081394.1 | - | 7 - 8 | 656 | 80 | |
| BC096969.1 | - | 5 - 6 | 80 | |||
| BC096970.1 | - | 6 - 7 | 80 | |||
More Information
Set Membership: |
|

zgc:101731
Genomic Map