Assay Details
| Gene Symbol: daam1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030136.2 | NP_001025307.1 | 20 - 21 | 2553 | 65 | |
| XM_005160502.4 | XP_005160559.1 | 21 - 22 | 2760 | 65 | ||
| XM_005160503.4 | XP_005160560.1 | 21 - 22 | 2954 | 65 | ||
| XM_017352491.2 | XP_017207980.1 | 20 - 21 | 2924 | 65 | ||
| GenBank mRNA | BC163491.1 | - | 19 - 20 | 2556 | 65 | |
More Information
Set Membership: |
|

daam1b
Genomic Map