Assay Details
| Gene Symbol: daam1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030136.2 | NP_001025307.1 | 6 - 7 | 887 | 95 | |
| XM_005160502.4 | XP_005160559.1 | 7 - 8 | 1094 | 95 | ||
| XM_005160503.4 | XP_005160560.1 | 7 - 8 | 1288 | 95 | ||
| XM_017352491.2 | XP_017207980.1 | 7 - 8 | 1288 | 95 | ||
| GenBank mRNA | BC163491.1 | - | 6 - 7 | 920 | 95 | |
More Information
Set Membership: |
|

daam1b
Genomic Map