Assay Details
| Gene Symbol: plvapb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030244.3 | NP_001025415.3 | 1 - 2 | 616 | 144 | |
| GenBank mRNA | AY972159.1 | - | 1 - 2 | 616 | 144 | |
| BC091873.1 | - | 1 - 2 | 544 | 144 | ||
| BC117570.1 | - | 1 - 2 | 681 | 144 | ||
| BC165836.1 | - | 1 - 2 | 427 | 144 | ||
More Information
Set Membership: |
|

plvapb
Genomic Map