Assay Details
| Gene Symbol: plvapb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030244.3 | NP_001025415.3 | 3 - 4 | 1392 | 86 | |
| GenBank mRNA | BC091873.1 | - | 3 - 4 | 1320 | 86 | |
| BC165836.1 | - | 3 - 4 | 1203 | 86 | ||
More Information
Set Membership: |
|

plvapb
Genomic Map