Assay Details
| Gene Symbol: plvapb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030244.3 | NP_001025415.3 | 4 - 5 | 1477 | 71 | |
| GenBank mRNA | AY972159.1 | - | 4 - 5 | 1477 | 71 | |
| BC091873.1 | - | 4 - 5 | 1405 | 71 | ||
| BC117570.1 | - | 4 - 5 | 1542 | 71 | ||
More Information
Set Membership: |
|

plvapb
Genomic Map