Assay Details
| Gene Symbol: acsl1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031837.1 | NP_001027007.1 | 14 - 15 | 1431 | 121 | |
| XM_005168349.4 | XP_005168406.1 | 14 - 15 | 1681 | 121 | ||
| XM_005168350.4 | XP_005168407.1 | 14 - 15 | 1509 | 121 | ||
| GenBank mRNA | BC100000.1 | - | 14 - 15 | 1431 | 121 | |
| BC155225.1 | - | 14 - 15 | 1466 | 121 | ||
More Information
Set Membership: |
|

acsl1a
Genomic Map