Assay Details
| Gene Symbol: pacsin1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033728.1 | NP_001028900.1 | 6 - 7 | 898 | 85 | |
| XM_005166140.3 | XP_005166197.1 | 6 - 7 | 992 | 85 | ||
| XM_009302728.3 | XP_009301003.1 | 6 - 7 | 1215 | 85 | ||
| GenBank mRNA | BC081593.1 | - | 6 - 7 | 954 | 85 | |
| BC097107.1 | - | 6 - 7 | 898 | 85 | ||
| BC165817.1 | - | 5 - 6 | 826 | 85 | ||
More Information
Set Membership: |
|

pacsin1b
Genomic Map