Assay Details
| Gene Symbol: pacsin1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033728.1 | NP_001028900.1 | 8 - 9 | 1147 | 90 | |
| XM_005166140.3 | XP_005166197.1 | 8 - 9 | 1241 | 90 | ||
| XM_009302728.3 | XP_009301003.1 | 8 - 9 | 1464 | 90 | ||
| GenBank mRNA | BC081593.1 | - | 8 - 9 | 1203 | 90 | |
| BC097107.1 | - | 8 - 9 | 1147 | 90 | ||
| BC165817.1 | - | 7 - 8 | 1075 | 90 | ||
More Information
Set Membership: |
|

pacsin1b
Genomic Map