Assay Details
| Gene Symbol: snx24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033731.1 | NP_001028903.1 | 1 - 2 | 240 | 75 | |
| GenBank mRNA | BC097192.1 | - | 1 - 2 | 240 | 75 | |
More Information
Set Membership: |
|

snx24
Genomic Map