Assay Details
| Gene Symbol: itgb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034971.1 | NP_001030143.1 | 13 - 14 | 2170 | 84 | |
| XM_005170612.4 | XP_005170669.1 | 14 - 15 | 2437 | 84 | ||
| XM_017353834.2 | XP_017209323.1 | 14 - 15 | 2437 | 84 | ||
| XM_017353835.2 | XP_017209324.1 | 14 - 15 | 2346 | 84 | ||
| GenBank mRNA | DQ149101.1 | - | 13 - 14 | 2170 | 84 | |
More Information
Set Membership: |
|

itgb1a
Genomic Map