Assay Details
| Gene Symbol: dusp23a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039620.1 | NP_001034709.1 | 1 - 2 | 407 | 58 | |
| GenBank mRNA | BC092774.1 | - | 1 - 2 | 334 | 58 | |
| BC109414.1 | - | 1 - 2 | 347 | 58 | ||
| BC121761.1 | - | 1 - 2 | 336 | 58 | ||
More Information
Set Membership: |
|

dusp23a
Genomic Map