Assay Details
| Gene Symbol: slc25a24l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021481188.1 | XP_021336863.1 | 4 - 5 | 721 | 102 | |
| GenBank mRNA | BC115225.1 | - | 4 - 5 | 696 | 102 | |
More Information
Set Membership: |
|

slc25a24l
Genomic Map