Assay Details
| Gene Symbol: pdlim3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042718.1 | NP_001036183.1 | 4 - 5 | 527 | 109 | |
| GenBank mRNA | BC163828.1 | - | 4 - 5 | 579 | 109 | |
| BC163837.1 | - | 4 - 5 | 579 | 109 | ||
| DQ679228.1 | - | 4 - 5 | 527 | 109 | ||
More Information
Set Membership: |
|

pdlim3b
Genomic Map