Assay Details
| Gene Symbol: aars1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044310.2 | NP_001037775.1 | 13 - 14 | 1859 | 70 | |
| XM_005159050.4 | XP_005159107.1 | 12 - 13 | 1788 | 70 | ||
| GenBank mRNA | BC071378.1 | - | 13 - 14 | 1864 | 70 | |
| BC128794.1 | - | 12 - 13 | 1745 | 70 | ||
More Information
Set Membership: |
|

aars1
Genomic Map