Assay Details
| Gene Symbol: asap2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044933.2 | NP_001038398.2 | 8 - 9 | 1003 | 63 | |
| XM_005160726.4 | XP_005160783.1 | 8 - 9 | 1046 | 63 | ||
| GenBank mRNA | BC129209.1 | - | 8 - 9 | 1003 | 63 | |
More Information
Set Membership: |
|

asap2b
Genomic Map