Assay Details
| Gene Symbol: pstpip1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045216.1 | NP_001038681.1 | 10 - 11 | 740 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pstpip1a| Gene Symbol: pstpip1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045216.1 | NP_001038681.1 | 10 - 11 | 740 | 112 | |
Set Membership: |
|