Assay Details
| Gene Symbol: otud5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075110.1 | NP_001068578.1 | 1 - 2 | 765 | 89 | |
| GenBank mRNA | BC124533.1 | - | 1 - 2 | 765 | 89 | |
| BC155304.1 | - | 2 - 3 | 682 | 89 | ||
More Information
Set Membership: |
|

otud5a
Genomic Map