Assay Details
| Gene Symbol: grin1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076714.2 | NP_001070182.2 | 3 - 4 | 922 | 94 | |
| XM_005161061.4 | XP_005161118.1 | 3 - 4 | 936 | 94 | ||
| XM_005161068.4 | XP_005161125.1 | 3 - 4 | 935 | 94 | ||
| XM_005161069.4 | XP_005161126.1 | 3 - 4 | 934 | 94 | ||
| XM_009295216.3 | XP_009293491.1 | 3 - 4 | 559 | 94 | ||
| XM_021468911.1 | XP_021324586.1 | 3 - 4 | 936 | 94 | ||
| XM_021468913.1 | XP_021324588.1 | 3 - 4 | 721 | 94 | ||
| GenBank mRNA | BC124078.1 | - | 3 - 4 | 883 | 94 | |
| KM655722.1 | - | 3 - 4 | 559 | 94 | ||
More Information
Set Membership: |
|

grin1a
Genomic Map