Assay Details
| Gene Symbol: ppm1h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077455.1 | NP_001070923.1 | 7 - 8 | 1140 | 59 | |
| GenBank mRNA | BC124421.1 | - | 7 - 8 | 1140 | 59 | |
More Information
Set Membership: |
|

ppm1h
Genomic Map