Assay Details
| Gene Symbol: slc39a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080619.1 | NP_001074088.1 | 448 | 65 | ||
| GenBank mRNA | BC129206.1 | - | 448 | 65 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

slc39a3| Gene Symbol: slc39a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080619.1 | NP_001074088.1 | 448 | 65 | ||
| GenBank mRNA | BC129206.1 | - | 448 | 65 | ||
Set Membership: |
|