Assay Details
| Gene Symbol: adat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082887.1 | NP_001076356.1 | 3 - 4 | 388 | 75 | |
| XM_009298175.2 | XP_009296450.1 | 3 - 4 | 441 | 75 | ||
| GenBank mRNA | BC133939.1 | - | 3 - 4 | 388 | 75 | |
More Information
Set Membership: |
|

adat1
Genomic Map