Assay Details
| Gene Symbol: clcn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082910.1 | NP_001076379.1 | 18 - 19 | 2125 | 93 | |
| XM_009294060.3 | XP_009292335.1 | 18 - 19 | 2456 | 93 | ||
| XM_017352282.2 | XP_017207771.1 | 17 - 18 | 2327 | 93 | ||
| XM_017352285.2 | XP_017207774.1 | 10 - 11 | 1232 | 93 | ||
| XM_021468144.1 | XP_021323819.1 | 18 - 19 | 2228 | 93 | ||
More Information
Set Membership: |
|

clcn1a
Genomic Map