Assay Details
| Gene Symbol: clcn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082910.1 | NP_001076379.1 | 19 - 20 | 2215 | 69 | |
| XM_009294060.3 | XP_009292335.1 | 19 - 20 | 2546 | 69 | ||
| XM_017352282.2 | XP_017207771.1 | 18 - 19 | 2417 | 69 | ||
| XM_017352285.2 | XP_017207774.1 | 11 - 12 | 1322 | 69 | ||
| XM_021468144.1 | XP_021323819.1 | 19 - 20 | 2318 | 69 | ||
More Information
Set Membership: |
|

clcn1a
Genomic Map