Assay Details
| Gene Symbol: clcn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082910.1 | NP_001076379.1 | 8 - 9 | 889 | 91 | |
| XM_009294060.3 | XP_009292335.1 | 8 - 9 | 1220 | 91 | ||
| XM_017352282.2 | XP_017207771.1 | 7 - 8 | 1091 | 91 | ||
| XM_021468144.1 | XP_021323819.1 | 8 - 9 | 992 | 91 | ||
More Information
Set Membership: |
|

clcn1a
Genomic Map