Assay Details
| Gene Symbol: klc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083550.1 | NP_001077019.1 | 608 | 62 | ||
| GenBank mRNA | BC134949.1 | - | 608 | 62 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

klc2| Gene Symbol: klc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083550.1 | NP_001077019.1 | 608 | 62 | ||
| GenBank mRNA | BC134949.1 | - | 608 | 62 | ||
Set Membership: |
|