Assay Details
| Gene Symbol: hip1rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083565.2 | NP_001077034.2 | 7 - 8 | 784 | 95 | |
| GenBank mRNA | BC133158.1 | - | 7 - 8 | 783 | 95 | |
More Information
Set Membership: |
|

hip1rb
Genomic Map